missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Antibody (Cureab patent anti-GP73) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28374JF585
This item is not returnable.
View return policy
Description
GOLM1 Monoclonal antibody specifically detects GOLM1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| GOLM1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| GOLM1 | |
| 0.1 mL | |
| Golgi Apparatus Markers, Signal Transduction | |
| 51280 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Cureab patent anti-GP73 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction