Golgin-97 Mouse anti-Human, eFluor 660, Clone: CDF4, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 50-9767-82

Code: NEW

Additional Details:
Additional Details: Weight: 0.09500kg



Disclaimers: For Research Use Only.

Product Code. 15550127

Quantity Price
1 £ 164.47 / 100µg
Estimated Shipment date
from Supplier 09-05-2017
Add to basket


Description and Specification

Specification

Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Conjugate eFluor 660
Gene Alias Golgin Subfamily A Member 1, Golgin A1
Monoclonal or Polyclonal Monoclonal
Isotype IgG1
Antigen Golgin-97
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 100μg
Regulatory Status RUO
Clone CDF4
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Concentration 0.2mg/mL
Host Species Mouse
Format Conjugated

The monoclonal antibody CDF4 recognizes human Golgin-97. Golgin-97, a GRIP domain protein member that was first identified as a Golgi complex autoantigen associated with Sjogren’s syndrome. Golgin-97 is a peripheral membrane protein found on the trans face of the Golgi apparatus. The protein G family member, Arl1, mediates the binding of Golgin-97 to the Golgi membrane. Like all GRIP proteins, Golgin-97 functions in maintaining the integrity of the trans Golgi network. Golgin-97 has also been shown to function as a tethering molecule in the retrograde transport of proteins from the endosomes to the trans Golgi network. During poxvirus infection, Golgin-97 incorporates into virions and plays a role in viral replication and morphogenesis.