missing translation for 'onlineSavingsMsg'
Learn More

Golgin-97 Mouse anti-Human, Alexa Fluor 488, Clone: CDF4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-9767-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15580287

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The monoclonal antibody CDF4 recognizes human Golgin-97. Golgin-97, a GRIP domain protein member that was first identified as a Golgi complex autoantigen associated with Sjogren’s syndrome. Golgin-97 is a peripheral membrane protein found on the trans face of the Golgi apparatus. The protein G family member, Arl1, mediates the binding of Golgin-97 to the Golgi membrane. Like all GRIP proteins, Golgin-97 functions in maintaining the integrity of the trans Golgi network. Golgin-97 has also been shown to function as a tethering molecule in the retrograde transport of proteins from the endosomes to the trans Golgi network. During poxvirus infection, Golgin-97 incorporates into virions and plays a role in viral replication and morphogenesis.
Specifications

Specifications

Golgin-97
CDF4
Alexa Fluor 488
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Immunocytochemistry, Immunohistochemistry, Microscopy
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Golgin Subfamily A Member 1, Golgin A1
IgG1, kappa
RUO
Primary
Human
Videos
Documents

For Research Use Only.