missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLGA6L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549314
Additional Details : Weight : 0.01000kg
Description
GOLGA6L2 Polyclonal antibody specifically detects GOLGA6L2 in Human samples. It is validated for Western Blot.Specifications
GOLGA6L2 | |
Polyclonal | |
Unconjugated | |
GOLGA6L2 | |
cancer/testis antigen 105; CT105; GOLGA6L2; golgi autoantigen, golgin subfamily a, 6-like 2; golgin A6 family-like 2; golgin subfamily A member 6-like protein 2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
283685 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q8N9W4 | |
GOLGA6L2 | |
KLH conjugated synthetic peptide between 104-130 amino acids from the N-terminal region of human GOLGA6L2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |