Learn More
Gold wire, 14kt, red, 1.63mm (0.064 in.) dia., Au 58.3% min, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 040328.BW
Code : NEW
Additional Details : CAS Number : 7440-57-5 Weight : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Red | |
1.63mm (0.064 in.) dia. | |
50 cm | |
% Concentration Cu>Ag, Au ≤58.3% | |
Wire |
Gold wire (14kt) | |
MFCD00003436 | |
Insoluble in water. | |
≈28g/m |