Learn More
Gold standard solution, for AAS, 1mg/mL Au in 0.5N HCl, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 196005000
131.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 29
Additional Details : CAS Number : 7732-18-5 Weight : 0.54500kg Transport : UN number : 1789 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Gold standard solution | |
1 mg/ml Au in 0.5N HCl | |
Plastic bottle | |
Orange/Yellow to Yellow | |
MFCD00003436 | |
[Au] | |
196.97 | |
CHEBI:30050 | |
AAS | |
1.09 | |
Powder and/or Chunks |
990 to 1010μg/mL of Au(3+) in 10 to 20% HCl | |
7440-57-5,7647-01-0,7732-18-5 | |
500 mL | |
Au | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
196.97 | |
1.0900g/mL | |
Soluble in water |
Safety and Handling
- Gold standard solution, 1 mg/ml Au in 10-20% HCl
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid
RUO – Research Use Only