missing translation for 'onlineSavingsMsg'
Learn More

Gold slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Premion™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number 15464775
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
This item is not returnable. View return policy
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]

Electronics, vapor deposition

Electronics, vapor depositionGold slug is used in thermal evaporation and electrodes (quartz oscillator, light-emitting diodes, and laser diodes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 Pc.
Solubility Information Soluble in hot sulfuric acid and aqua regia. Insoluble in water and acid.
Percent Purity 99.995%
Grade Premion™
Odor Odorless
Weight ≈0.49g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Gold slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'