missing translation for 'onlineSavingsMsg'
Learn More

Gold powder, -100 mesh, Premion™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number 15436625
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
This item is not returnable. View return policy
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]

Gold, Au, CAS Number-7440-57-5, burnish, leaf, c.i. pigment metal 3, flake, magnesium purple, colloidal, ci pigment metal 3, shell, powder, gold, colloidal, 5g, Gold colored powder, 99.998% (Metals basis), CHEBI:30050, Golden, 19.32g/cm3 at 20 deg.C, 231-165-9, PCHJSUWPFVWCPO-UHFFFAOYSA-N, -100 Mesh

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Golden
Appearance Gold colored powder
Assay Percent Range 99.998% (Metals basis)
Quantity 1 g
Mesh Size -100 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Gold

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'