missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold plating solution, electroless, metal content≈3.7g/L

For coating nickel or copper

Brand:  Alfa Aesar™ 042307.AK

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 2922 Chem class : 1 Pack group : III

Quantity 250mL

 View more versions of this product

Product Code. 11324358

  • £123.00 / 250mL


Estimated Shipment date
from Supplier 18-01-2018
Add to basket

Specifications

Chemical Identifiers

Gold plating solution
Electroless, metal content ≈3.7g/L
MFCD00003436
UN2922

Specifications

Gold plating solution
Liquid
UN2922
250mL
Electroless, metal content ≈3.7g/L
MFCD00003436
Colorless to pale yellow
Videos
SDS