missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold Phosphorus slug, 3.175mm (0.125 in.) dia. x 9.525mm (0.375 in.) length, Premion™, 99.99% (metals basis)

Brand:  Alfa Aesar™ 039464.KJ

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 5 pieces

 View more versions of this product

Product Code. 15443705

  • £579.50 / Pack of 5


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Gold Phosphorus slug
3.175mm (0.125 in.) dia. x 9.525mm (0.375 in.) length
Premion™
MFCD00003436

Specifications

Au:P; 99.5:0.5 wt%
5 pieces
Premion™
99.99%
MFCD00003436
Gold Phosphorus slug
3.175mm (0.125 in.) dia. x 9.525mm (0.375 in.) length
Slug
(metals basis)
Videos
SDS