Learn More
Thermo Scientific Alfa Aesar Gold nanorods, 19-25nm dia. x 63-73nm length, dispersion in water, 700nm Absorption, negatively charged
Brand:  Thermo Scientific Alfa Aesar 046923.AA
Gold nanorods, 19-25nm dia. x 63-73nm length, dispersion in water, 700nm Absorption, negatively charged
Code : NEW
Additional Details : CAS Number : 7440-57-5 Weight : 0.00005kg
Product Code. 15485045