missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold nanorods, 19-25nm dia. x 63-73nm length, dispersion in water, 700nm Absorption, negatively charged

Brand:  Alfa Aesar™ 046923.AC

Code : NEW

Additional Details : CAS Number : 7440-57-5 Weight : 0.00025kg

Quantity 25mL

 View more versions of this product

Product Code. 15495045

  • £160.06 / 25mL


Estimated Shipment date
from Supplier 14-05-2018
Add to basket

Specifications

Chemical Identifiers

Gold nanorods
7440-57-5
19 to 25nm dia. x 63 to 73nm length, dispersion in water, 700nm Absorption, negatively charged
MFCD00003436

Specifications

Gold nanorods
19 to 25nm dia. x 63 to 73nm length, dispersion in water, 700nm Absorption, negatively charged
Liquid
Odorless
>60°C (dec.)
25mL
7440-57-5
MFCD00003436
Colorless
Videos
SDS