missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 90nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 564nm absorption, Thermo Scientific™

Catalog Number 15444545
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
This item is not returnable. View return policy
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
TRUSTED_SUSTAINABILITY
Color Pink to Red
Quantity 100 mL
Particle Size 90nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 564nm absorption
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'