missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 5nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 515-520nm absorption, Thermo Scientific Chemicals

Catalog Number 15474635
Click to view available options
Quantity:
25 mL
100 mL
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]

Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red
Quantity 25 mL
Percent Purity 99%
Odor Odorless
Particle Size 5nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, supplied in 0.1mM PBS, reactant to free, 99%, OD1, 515 to 520nm absorption

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'