missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold nanoparticles, 50nm, supplied in 0.1mM PBS, 95%, OD1, 535nm absorption

Brand:  Alfa Aesar™ J67157.AE

Code : NEW

Additional Details : Weight : 0.00010kg

Quantity 100mL

 View more versions of this product

Product Code. 15494535

  • £131.60 / 100mL


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Description

Description

  • For conjugate development by passive adsorption of proteins and other ligands to the gold nanoparticle surface
  • Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications
Specifications

Chemical Identifiers

Gold nanoparticles
7440-57-5
supplied in 0.1mM PBS, 95%, OD: 1, 535nm absorption
MFCD00003436

Specifications

Gold nanoparticles
supplied in 0.1mM PBS, 95%, OD: 1, 535nm absorption
Liquid
MFCD00003436
Red to pink
100mL
7440-57-5
95%
50nm
Videos
SDS