missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 50nm, supplied in 0.1 mg/ml sodium citrate with stabilizer OD1, 535nm absorption, Thermo Scientific Chemicals

Catalog Number 15404635
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
This item is not returnable. View return policy
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]

For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
TRUSTED_SUSTAINABILITY
Color Pink to Red
Quantity 25 mL
Particle Size 50nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer OD: 1, 535nm absorption

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'