missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
Description
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown to Pink |
Quantity | 100 mL |
Percent Purity | 99% |
Particle Size | 250nm |
Sensitivity | Light sensitive |
Form | Liquid |
Chemical Name or Material | Gold nanoparticles, supplied in 0.1mM PBS, reactant-free |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'