Learn More
Thermo Scientific Alfa Aesar Gold nanoparticles, 20nm, Transferrin conjugated, OD3, supplied in 0.01M PBS (pH 7.4), 20% glycerol v/v, 1% BSA
Brand:  Thermo Scientific Alfa Aesar J67165.XF
Gold nanoparticles, 20nm, Transferrin conjugated, OD3, supplied in 0.01M PBS (pH 7.4), 20% glycerol v/v, 1% BSA
Code : NEW
Additional Details : Weight : 0.00001kg
Product Code. 15404545