missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold nanoparticles, 150nm, supplied in 0.1mg/mL sodium citrate with stabilizer

Brand:  Alfa Aesar™ J67342.AC

Code : NEW

Additional Details : Weight : 0.00025kg

Quantity 25mL

 View more versions of this product

Product Code. 15494595

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Gold nanoparticles
7440-57-5
supplied in 0.1mg/mL sodium citrate with stabilizer
MFCD00003436

Specifications

Gold nanoparticles
supplied in 0.1mg/mL sodium citrate with stabilizer
Liquid
150nm
25mL
7440-57-5
MFCD00003436
Dark brown to pink
Videos
SDS