missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold nanoparticles, 150nm, supplied in 0.1mg/mL sodium citrate with stabilizer

Brand:  Alfa Aesar™ J67342.AE

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 0.00010kg

Quantity 100mL

 View more versions of this product

Product Code. 15404605

  • £163.24 / 100mL


Estimated Shipment date
from Supplier 04-09-2017
Add to basket

Specifications

Chemical Identifiers

Gold nanoparticles
7440-57-5
supplied in 0.1mg/mL sodium citrate with stabilizer
MFCD00003436

Specifications

Gold nanoparticles
supplied in 0.1mg/mL sodium citrate with stabilizer
Liquid
150nm
100mL
7440-57-5
MFCD00003436
Dark brown to pink
Documentation

Technical Documents