missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride, Premion™, 99.99% (metals basis), Au 64.4% min, Thermo Scientific Chemicals

Catalog Number p-4407503
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13453-07-1
AuCl3
MFCD00014172
This item is not returnable. View return policy
13453-07-1
AuCl3
MFCD00014172

Gold(III) chloride is used in colloidal gold solutions, in photography and as a print toning agent(gold toning), starting solution to form other gold compounds and a precursor for preparation of ultra pure gold metal. It is used in electroplating and electroless plating as an anode in an electric cell. Gold(III) chloride acts as a gold catalyst and cell body stains for bright field and dark field microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 180° (sublimation)
Color Red
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water,ethanol,ether and slightly soluble in liquid ammonia.
Formula Weight 303.33
Odor Odorless
Sensitivity Hygroscopic
Appearance Red crystalline
Density 3.90 g/cm3
Chemical Name or Material Gold(III) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'