Learn More
Thermo Scientific Alfa Aesar Gold(I) sodium cyanide, 99.9% (metals basis), Au 72% min
Brand: Thermo Scientific Alfa Aesar 012637.01
Gold(I) sodium cyanide, 99.9% (metals basis), Au 72% min
Code : NEW
Additional Details : CAS Number : 15280-09-8 Weight : 0.00025kg Transport : UN number : 1588 Chem class : 6.1 Pack group : II