missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold(I) sodium cyanide, 99.9% (metals basis), Au 72% min

Brand:  Alfa Aesar™ 012637.06

Code : NEW

Additional Details : CAS Number : 15280-09-8 Weight : 0.00500kg Transport : UN number : 1588 Chem class : 1 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11307487

  • £614.80 / 5g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

Gold(I) sodium cyanide
NaAu(CN)2
MFCD00050423
14,8610
15280-09-8
271.99
UN1588
Sodium gold cyanide; Sodium dicyanoaurate(I)

Specifications

Gold(I) sodium cyanide
Powder
(metals basis)
271.99
UN1588
Sodium gold cyanide; Sodium dicyanoaurate(I)
Characteristic
15280-09-8
99.9%
NaAu(CN)2
MFCD00050423
14,8610
Au 72% min.
5g
Videos
SDS