missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) cyanide, Yellow powder, Premion™, 99.99% (Metals basis), Alfa Aesar™

Brand:  Alfa Aesar 032031.01

Code : NEW

Additional Details : CAS Number : 506-65-0 Weight : 0.00025kg Transport : UN number : 1588 Chem class : 1 Pack group : II

Quantity 0.25g

 View more versions of this product

Product Code. 11318317

  • £50.14 / 0.25g


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Chemical Identifiers

Gold(I) cyanide
Powder
AuCN
MFCD00003437
506-65-0
99.99% (Metals basis)
222.98

Specifications

Gold(I) cyanide
Powder
AuCN
MFCD00003437
Moisture sensitive
0.25g
506-65-0
99.99% (Metals basis)
222.98
Almond-like
Yellow
Videos
SDS