missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) cyanide, Yellow powder, Premion™, 99.99% (Metals basis), Alfa Aesar™

Brand:  Alfa Aesar 032031.06

Code : NEW

Additional Details : CAS Number : 506-65-0 Weight : 0.00500kg Transport : UN number : 1588 Chem class : 6.1 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11328317

  • £552.37 / 5g


Estimated Shipment date
from Supplier 25-06-2018
Add to basket

Specifications

Chemical Identifiers

Gold(I) cyanide
Powder
AuCN
MFCD00003437
506-65-0
99.99% (Metals basis)
222.98

Specifications

Gold(I) cyanide
Powder
AuCN
MFCD00003437
Moisture sensitive
5g
506-65-0
99.99% (Metals basis)
222.98
Almond-like
Yellow
Videos
SDS