missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) cyanide, ACROS Organics™

Brand:  Acros Organics 390132500

Code : 29

Additional Details : CAS Number : 506-65-0 Weight : 0.05025kg Transport : UN number : 1588 Chem class : 6.1 Pack group : II


Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10541524

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Gold(I) cyanide
AuCN
222.98
506-65-0
AuCN
MFCD00003437

Specifications

Gold(I) cyanide
Powder
AuCN
MFCD00003437
87.0 min.
Solubility in water: negligible. Other solubilities: practically insoluble in alcohol, ether and, diluted acids, soluble in ammonia, solutions of nacn
250mg
506-65-0
AuCN
222.98
15, 4551
Glass bottle
Yellow
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Gold(I) cyanide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P361-Take off immediately all contaminated clothing.

SDS