missing translation for 'onlineSavingsMsg'
Learn More

Gold Germanium slug, 6.35mm (0.25in)dia x 6.35mm (0.25 in.) length, Premion™, 99.999% (metals basis), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 014173.KJ

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15493495

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Gold Germanium slug
Au:Ge; 88:12 wt%
MFCD02091706
99.999%
Odorless
Slug
6.35mm (0.25 in.)dia. x 6.35mm (0.25 in.) length
(metals basis)
5 Pc.
Premion™
≈2.9g/slug
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers