missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold Beryllium pieces, 99.9% (metals basis)

Brand:  Alfa Aesar™ 045489.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15482297

  • £144.48 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Gold Beryllium pieces
MFCD00003436

Specifications

Au:Be; 99:1 wt%
1g
(metals basis)
Odorless
Gold Beryllium pieces
99.9%
MFCD00003436
3.175mm to 6.35mm
Videos
SDS