missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold Beryllium pieces, 99.9% (metals basis)

Brand:  Alfa Aesar™ 045489.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15492297

  • £578.02 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Gold Beryllium pieces
MFCD00003436

Specifications

Au:Be; 99:1 wt%
5g
(metals basis)
Odorless
Gold Beryllium pieces
99.9%
MFCD00003436
3.175mm to 6.35mm
Videos
SDS