missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold Beryllium pellet, 2.00mm (0.08 in.) dia. x 10mm (0.39 in.) length, slightly pitted surface

Brand:  Alfa Aesar™ 045608.KU

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 5 each

 View more versions of this product

Product Code. 15464005

  • £331.10 / Pack of 5


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

Gold Beryllium pellet
2.00mm (0.08 in.) dia. x 10mm (0.39 in.) length, slightly pitted surface
MFCD00003436

Specifications

Gold Beryllium pellet
Pellet
Au:Be; 99:1 wt%
5 each
2.00mm (0.08 in.) dia. x 10mm (0.39 in.) length, slightly pitted surface
MFCD00003436
Odorless
≈0.554g/pellet
Videos
SDS