missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ GoggleGear™ 500 Series Prescription Insert
7.90 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Prescription Insert
Description
- Insert is coated with Scotchgard™ Anti-Fog coating which retains its effectiveness for at least 25 washings with water, allowing workers to wear their eyewear longer.
- 3M Catalog# GG500-PI
Specifications
Color | Clear |
Product Type | Prescription insert |
For Use With (Equipment) | 3M Goggle Gear, 500-Series Splash Goggles |
Model | GG500-PI |
Material | Polycarbonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'