Learn More
F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, PE, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen 31864
Code : PN
Additional Details : Weight : 0.00100kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31864 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31864 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, mouse, and rat serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Phycobiliprotein Concentration: 0.5 mg/mL (determined by absorption = 82.0 at 566 nm for a 1% solution for only those R-PE molecules to which at least one molecule of active antibody is bound) Phycoerythrin Source: Seaweed Store product protected from light at 4°C until opened. DO NOT FREEZE. RPE Amax= 490 nm, 545 nm, and 566 nm; Emax= 580 nm. Reconstitute with 1.0 mL of distilled water (0.5 mg/mL after restoration). Country of Origin: USA
Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Flow Cytometry.Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
PE | |
Goat | |
1 mL | |
Secondary | |
4° C, do not freeze | |
IgG |
Flow Cytometry | |
0.5 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Rabbit | |
Lyophilized |