Goat anti-Rabbit IgG, (H+L) Affinity Purified F(ab')2, Secondary Antibody Millipore™ (Chemicon™)


Overview
Brand: Merck Millipore

Manufacturer Part Number: AQ132P

UNSPSC: 12352203

Code: LD

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 11347630

Quantity Price
1 £ 199.34 / 500µL
Estimated Shipment date
from Supplier 03-05-2017
Add to basket

Please note: We no longer supply Millipore Antibodies.


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Antigen Goat anti-Rabbit IgG, (H+L) Affinity Purified F(ab')2
Cross Reactivity Rabbit
Primary or Secondary Secondary
Format Semi-purified
Regulatory Status RUO
Host Species Goat
Conjugate HRP
Quantity 500μL
Monoclonal or Polyclonal Polyclonal
Alternative Products