Learn More
Goat anti-Mouse IgM Secondary Antibody, Biotin, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen 31804
107.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : PN
Additional Details : Weight : 0.80000kg
Description
Product # 31804 reacts with the μ chains of mouse IgM. This antibody cross reacts with Rat IgM (6%). It has minimal cross-reactivity with immunoglobulins from other species.
Mouse IgM Polyclonal antibody specifically detects Mouse IgM in Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot.Specifications
Mouse IgM | |
Polyclonal | |
Biotin | |
Goat | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. | |
IgG |
Immunocytochemistry, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot | |
0.5 mg/mL | |
PBS with 1mg/mL BSA and 0.08% sodium azide; pH 7.8 | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |