missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG2c Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA128887
142.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-28887 detects Mouse IgG2c from mouse samples.
Mouse IgG2c Polyclonal antibody specifically detects Mouse IgG2c in Mouse samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Specifications
Mouse IgG2c | |
Polyclonal | |
Unconjugated | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Mouse | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goats were immunized with highly purified IgG2c isolated from C57BL/6 mice, in Freund’s adjuvant. | |
500 μg | |
Secondary | |
4° C | |
IgG |