Learn More
IgG2c Goat anti-Mouse, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA129288
Code : LE
Additional Details : Weight : 1.24740kg
Description
PA1-29288 detects Mouse IgG2c from mouse samples. PA1-29288 has been successfully used in Western blot and ELISA applications. The PA1-29288 immunogen is: Goats were immunized with highly purified IgG 2c isolated from C57BL/6 mice, in Freunds adjuvant.
Thermo Scientific Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgG2c | |
Polyclonal | |
HRP | |
Goats were immunized with highly purified IgG 2c isolated from C57BL/6 mice, in Freund′s adjuvant. | |
500 μg | |
Secondary | |
4° C, do not freeze | |
IgG |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Goat | |
Purified | |
RUO | |
Mouse | |
Liquid |