Learn More
Goat anti-Mouse IgG, IgM (H+L) Cross-Adsorbed Secondary Antibody, AP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen 31330
Code : PN
Additional Details : Weight : 0.00100kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. The antibody has been successfully used in Western blot and IP applications. Antibody Specificity: This antibody reacts with the heavy chains of mouse IgG and IgM, and with the light chains common to most mouse immunoglobulins, based on immunoelectrophoresis. No antibody was detected against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reactivity with human, bovine and horse serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 1.0 mL distilled water (0.6 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA
Mouse IgG, IgM (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG, IgM (H+L) Cross-Adsorbed in Mouse samples. It is validated for Western Blot.Specifications
Mouse IgG, IgM (H+L) Cross-Adsorbed | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Goat | |
1 mL | |
Secondary | |
4° C | |
IgG |
Western Blot | |
0.6 mg/mL | |
TBS with 15mg/mL BSA and 0.05% sodium azide; pH 8 | |
Affinity chromatography | |
RUO | |
Mouse | |
Lyophilized |