Learn More
Goat anti-Human IgA Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA174395
148.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LE
Additional Details : Weight : 1.24740kg
Description
Goat anti human IgA polyclonal antibody recognizes the heavy chain of human IgA. This anti human IgA antibody has been cross adsorbed against human IgM and IgG but might cross-react with IgA from other species.
Human IgA Polyclonal antibody specifically detects Human IgA in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
Human IgA | |
Polyclonal | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.4 | |
Human IgA paraproteins. | |
1 mL | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |