Learn More
GPSM2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GPSM2.
Brand: Abnova PAB7195.100ug
Additional Details : Weight : 0.02000kg
Description
Heterotrimeric G proteins transduce extracellular signals received by cell surface receptors into integrated cellular responses. GPSM2 belongs to a group of proteins that modulate activation of G proteins (Blumer et al., 2002 [PubMed 11832491]).[supplied by OMIM
Sequence: C-NRLKGKKYKTNSSTKSpecifications
GPSM2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:64000) Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user. | |
GPSM2 | |
LGN/Pins | |
Goat | |
Antigen affinity purification | |
RUO | |
29899 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of GPSM2. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_0.37428.2 | |
GPSM2 | |
A synthetic peptide corresponding to human GPSM2. | |
100 μg | |
Primary | |
Human | |
Liquid |
For Research Use Only