missing translation for 'onlineSavingsMsg'
Learn More

Go 6983, Tocris Bioscience™

Broad spectrum PKC inhibitor

Brand:  Tocris (Bio-Techne) 228510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13430944

  • £246.96 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Broad spectrum protein kinase C (PKC) inhibitor (IC50 values are 7, 7, 6, 10, 60 and 20000 nM for PKCα, PKCβ, PKCγ, PKCδ, PKCζ and PKCμ respectively). Displays cardioprotective properties; reduces polymorphonuclear leukocyte adherence and infiltration following myocardial ischemia/reperfusion injury. Optimizes naïve human pluripotent stem cell growth and viability following naïve cell derivation from primed ESCs and iPSCs using naïve human stem cell medium (NHSM).
Specifications

Chemical Identifiers

133053-19-7
442.519
gö 6983, 3-1-3-dimethylamino propyl-5-methoxy-1h-indol-3-yl-4-1h-indol-3-yl-1h-pyrrole-2,5-dione, 2-1-3-dimethylaminopropyl-5-methoxyindol-3-yl-3-1h-indol-3-yl maleimide, gouml 6983, d06tll, go, g, 3-1-3-dimethylamino propyl-5-methoxyindol-3-yl-4-1h-indol-3-yl pyrrole-2,5-dione
3-[1-[3-(dimethylamino)propyl]-5-methoxyindol-3-yl]-4-(1H-indol-3-yl)pyrrole-2,5-dione
C26H26N4O3
LLJJDLHGZUOMQP-UHFFFAOYSA-N
3499
CN(C)CCCN1C=C(C2=C1C=CC(=C2)OC)C3=C(C(=O)NC3=O)C4=CNC5=CC=CC=C54

Specifications

Go 6983
133053-19-7
gö 6983, 3-1-3-dimethylamino propyl-5-methoxy-1h-indol-3-yl-4-1h-indol-3-yl-1h-pyrrole-2,5-dione, 2-1-3-dimethylaminopropyl-5-methoxyindol-3-yl-3-1h-indol-3-yl maleimide, gouml 6983, d06tll, go, g, 3-1-3-dimethylamino propyl-5-methoxyindol-3-yl-4-1h-indol-3-yl pyrrole-2,5-dione
CN(C)CCCN1C=C(C2=C1C=CC(=C2)OC)C3=C(C(=O)NC3=O)C4=CNC5=CC=CC=C54
442.519
442.51
10mg
C26H26N4O3
LLJJDLHGZUOMQP-UHFFFAOYSA-N
3-[1-[3-(dimethylamino)propyl]-5-methoxyindol-3-yl]-4-(1H-indol-3-yl)pyrrole-2,5-dione
3499
>98%
Videos
Documents