Learn More
GNL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555529
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RIPFFVKPPN AEPLVAPQLL PSSSLEVVPE AAQNNPGEEV TETAGEGSES IIKEETEENS HCDANTEMQQ ILTRVRQNFG KINVVPQFSG DDLVPV Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 68%.
GNL2 Polyclonal antibody specifically detects GNL2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GNL2 | |
Polyclonal | |
Unconjugated | |
GNL2 | |
Autoantigen NGP-1; BC003262; G protein nucleolar 2; GNL2; guanine nucleotide binding protein-like 2 (nucleolar); HUMAUANTIG; NGP1; Ngp-1; Nog2; novel nucleolar guanosine 5'-triphosphate binding protein autoantigen; nucleolar G-protein gene 1; nucleolar GTPase; nucleolar GTP-binding protein 2; nucleostemin-2; Nug2; RGD1305006; testicular antigen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29889 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13823 | |
GNL2 | |
Recombinant protein corresponding to Human GNL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only