missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNG11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560963
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EQLRKEVKLQ RQQVSKCSEE IKNY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
GNG11 Polyclonal antibody specifically detects GNG11 in Human samples. It is validated for Immunohistochemistry.Specifications
GNG11 | |
Polyclonal | |
Unconjugated | |
GNG11 | |
0610037B21Rik; G protein gamma-11 subunit; G protein subunit gamma 11; GNG11; GNGT11; guanine nucleotide binding protein (G protein), gamma 11; guanine nucleotide binding protein gamma subunit 11; guanine nucleotide-binding protein G(I)/G(S)/G(O) gamma-11 subunit; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2791 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P61952 | |
GNG11 | |
Recombinant protein corresponding to Human GNG11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only