Learn More
GM-CSF Monoclonal Antibody (MP1-22E9), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-7331-81
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The MP1-22E9 antibody reacts with mouse granulocyte/macrophage - colony stimulating factor (GM-CSF). The MP1-22E9 antibody is a neutralizing antibody. Mouse GM-CSF is a 14 kDa factor produced mainly by activated T cells and macrophages. Other cell types, such as endothelium and fibroblasts, also secrete GM-CSF in response to TNF-alpha, IL-2, IL-1, and IFN-gamma. GM-CSF stimulates growth of macrophages, granulocytes and dendritic cells. GM-CSF is found as a membrane-bound form and also as a complex associated with the extracellular matrix. Non-glycosylated GM-CSF is biologically active. Applications Reported: The MP1-22E9 antibody has been reported for use in ELISA, ELISPOT, neutralization, and intracellular staining for flow cytometric analysis. Applications Tested: The Functional Grade Purified MP1-22E9 antibody has been tested by LAL assay to verify low endotoxin levels, by ELISA, and in bioassay for neutralization of GM-CSF bioactivity. The MP1-22E9 antibody at 1.25 μg/mL has been found to inhibit by 50% the biological effects of 1 ng/mL mouse GM-CSF in an MC/9 cell proliferation assay. Detailed information and protocols about cytokine bioassays and in vitro cytokine neutralization using antibodies can be found in the BestProtocols™ section.
The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.Specifications
GM-CSF | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P01587 | |
CSF2 | |
Affinity chromatography | |
RUO | |
12981 | |
4° C | |
Liquid |
ELISA, Functional Assay, Neutralization | |
MP1-22E9 | |
Functional Grade | |
CSF2 | |
colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim | |
Rat | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.