missing translation for 'onlineSavingsMsg'
Learn More

GM-CSF, Biotin, clone: MP1-31G6, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-7332-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15390060

  • £176.67 / 500µg


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The MP1-31G6 antibody reacts with mouse granulocyte/macrophage - colony stimulating factor (GM-CSF), a 14kDa cytokine secreted by activated T cells and macrophages.Applications Reported: MP1-31G6 has been reported for use in detection of mouse GM-CSF by ELISA.Applications Tested: The MP1-31G6 antibody has been tested as the detection antibody in a sandwich ELISA for analysis of mouse GM-CSF (Granulocyte/Macrophage-Colony Stimulating Factor, GMCSF) in combination with the purified MP1-22E9 (14-7331) antibody for capture and recombinant mouse GM-CSF (14-8331) as the standard. A suitable range of concentrations of this antibody for ELISA detection is 0.5-2 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

GM-CSF
MP1-31G6
Biotin
Conjugated
Rat
500μg
Store at 2-8°C. Do not freeze.
Monoclonal
ELISPOT Detection Antibody
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Granulocyte-macrophage colony stimulating factor, GMCSF, Colony-stimulating factor, CSF12
IgG1
RUO
Primary
Murine
Videos