missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine GM-CSF Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | GM-CSF |
---|---|
Clone | 249201 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Description
GM-CSF Monoclonal specifically detects GM-CSF in Canine samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
GM-CSF | |
LYOPH | |
ELISA | |
Unconjugated | |
Mouse | |
Canine | |
P48749.1 | |
1437 | |
E. coli-derived recombinant canine GM-CSF Ala18-Lys144 Accession # P48749.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects canine GM-CSF in ELISAs. In ELISAs, this antibody does not cross-react with recombinant human GM-CSF, recombinant mouse GM-CSF, recombinant rat GM-CSF, recombinant feline GM-CSF, recombinant porcine GM-CSF, or recombinant canine IL-5. |
249201 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
CSF2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only