missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ GM-CSF Recombinant Human Protein
Recombinant Human GM-CSF
Brand: Gibco™ CTP2011
Code : 50
Additional Details : Weight : 0.99000kg
Description
- Involved in many biological responses including growth and development of granulocyte and macrophage progenitor cells
- For stimulation and initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils
- ED50 range = 0.02 to 0.1 ng/mL, determined by dose dependent proliferation of human TF-1 cells
- Optimal concentration for individual application should be determined by dose response assay
Specifications
Cell Culture | |
Gel-Purified | |
Human | |
Human | |
Lyophilized protein can be stored at 2°C to 8°C dessicated. Once reconstituted, store in aliquots at ≤20°C. Avoid multiple freeze⁄thaw cycles. | |
0.055 ng/ml | |
<0.1 ng/μg | |
Wet Ice | |
Cytokines & Receptors | |
GM-CSF, G-CSF, & M-CSF | |
Gibco™ |
14 kDa | |
100 μg | |
GM-CSF | |
Recombinant | |
Carrier-Free | |
Proliferation of TF1 cells | |
Human | |
E. coli | |
Full Length, Recombinant, Ligand | |
Clinical Research, Oncology, Stem Cell Research, Differentiation, Signal Transduction, Drug Development | |
95 % |
For Research Use Only. Not for use in diagnostic procedures.