missing translation for 'onlineSavingsMsg'
Learn More

Glyoxalbis(2-hydroxyanil), 97%, Thermo Scientific Chemicals

Catalog Number p-3758070
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1149-16-2
C14H12N2O2
240.26
MFCD00002183
OMJNMRJMNUREOJ-KOBPNRPCSA-N
glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
5356911
OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1149-16-2
C14H12N2O2
240.26
MFCD00002183
OMJNMRJMNUREOJ-KOBPNRPCSA-N
glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
5356911
OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Melting Point 201°C to 205°C
Color Beige
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Solubility Information (0.5% in methanol) Clear
IUPAC Name 6-{[(1E)-2-[(2-hydroxyphenyl)amino]ethenyl]imino}cyclohexa-2,4-dien-1-one
Formula Weight 240.26
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material Glyoxalbis(2-hydroxyanil)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'