missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycylglycine, 99%

Catalog Number p-7080814
Click to view available options
Quantity:
25 g
100 g
500 g
556-50-3
C4H8N2O3
132.119
MFCD00008130
YMAWOPBAYDPSLA-UHFFFAOYSA-N
glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
11163
CHEBI:17201
2-[(2-aminoacetyl)amino]acetic acid
C(C(=O)NCC(=O)O)N
556-50-3
C4H8N2O3
132.119
MFCD00008130
YMAWOPBAYDPSLA-UHFFFAOYSA-N
glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
11163
CHEBI:17201
2-[(2-aminoacetyl)amino]acetic acid
C(C(=O)NCC(=O)O)N

Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.

Solubility
Soluble in water (132 g/L) at 20°C, and ethanol (slightly solube).

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼220°C (decomposition)
Beilstein 1765223
Merck Index 14,4503
Solubility Information Soluble in water (132g/L) at 20°C,and ethanol (slightly solube).
Formula Weight 132.12
Percent Purity 99%
Quantity 25 g
Chemical Name or Material Glycylglycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'