missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-Glutamic Acid, Fisher BioReagents

Catalog Number 10587714
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Amber Glass
This item is not returnable. View return policy
7412-78-4
C7H12N2O5
204.182
IEFJWDNGDZAYNZ-BYPYZUCNSA-N
glycyl-l-glutamic acid, n-glycylglutamic acid, gly-glu, n-glycyl-l-glutamic acid, s-2-2-aminoacetamido pentanedioic acid, 2s-2-2-aminoacetamido pentanedioic acid, l-glutamic acid, n-glycyl, glycyl-l-glutamicacid, gly-l-glu, gly-l-glu-oh
99278
CHEBI:73801
(2S)-2-[(2-aminoacetyl)amino]pentanedioic acid
C(CC(=O)O)C(C(=O)O)NC(=O)CN
This item is not returnable. View return policy
7412-78-4
C7H12N2O5
204.182
IEFJWDNGDZAYNZ-BYPYZUCNSA-N
glycyl-l-glutamic acid, n-glycylglutamic acid, gly-glu, n-glycyl-l-glutamic acid, s-2-2-aminoacetamido pentanedioic acid, 2s-2-2-aminoacetamido pentanedioic acid, l-glutamic acid, n-glycyl, glycyl-l-glutamicacid, gly-l-glu, gly-l-glu-oh
99278
CHEBI:73801
(2S)-2-[(2-aminoacetyl)amino]pentanedioic acid
C(CC(=O)O)C(C(=O)O)NC(=O)CN
Glycyl-L-Glutamic Acid is used in biochemical research applications.
TRUSTED_SUSTAINABILITY
Melting Point 155°C
Color Beige
Packaging Amber Glass
Quantity 1 g
Formula Weight 204.18
Physical Form Solid
Chemical Name or Material Glycyl-L-Glutamic Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'