missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Glycine-N,N,O-D3, 99.99% (Isotopic)

Brand:  Alfa Aesar™ H64075.MD

Code : NEW

Additional Details : CAS Number : 4896-76-8 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15411119

  • £31.38 / 250mg


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

Glycine-N,N,O-D3
Isotopic
78.09
4896-76-8
D2NCH2CO2D
MFCD00074933

Specifications

Glycine-N,N,O-D3
4896-76-8
99.99%
78.09
White
250mg
Isotopic
Crystalline Solid
D2NCH2CO2D
MFCD00074933
240°C (dec.)
Videos
SDS